missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Pop Beads

Catalog No. S000862
Change view
Click to view available options
Product Type:
1-hole Pop Bead
5-hole Pop Bead
Connector
Pop Beads
Color:
Black
Blue
Clear
Green
Lilac
Orange
Pink
Red
White
Yellow
Quantity:
10 Pk.
100 Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color Quantity
S000862 Pop Beads White 10 Pk.
S000849 1-hole Pop Bead Black 100 Pk.
S000878 1-hole Pop Bead Blue 100 Pk.
S000861 Connector Clear 100 Pk.
S000850 1-hole Pop Bead Green 100 Pk.
S000879 1-hole Pop Bead Lilac 100 Pk.
S000882 1-hole Pop Bead Orange 100 Pk.
S000876 1-hole Pop Bead Pink 100 Pk.
S000893 1-hole Pop Bead Red 100 Pk.
S000877 1-hole Pop Bead White 100 Pk.
S000894 5-hole Pop Bead White 100 Pk.
S000860 1-hole Pop Bead Yellow 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Choking Hazard
Catalog No. S000862 Supplier United Scientific™ Supplier No. UNCENTRM01
Choking Hazard

Pop beads are a useful tool to help visualize the small complex structure of biological molecules.

  • The beads connect easily and can be quickly disassembled to simulate different molecular activities.
  • With these hands-on models, students can visually and tactually feel DNA models and simulate mitosis and meiosis.
  • The specially designed white pop beads with four extra holes,equally spaced in a 90° plane from the axis of attachment, are typically used to form the nucleotide backbone. These are 10 mm diameter in size. The clear pop bead connectors are designed to simulate hydrogen bonds.
  • Various colors to make a wide variety of models and kits

Caution: Choking Hazard: Children under 8 years can choke or suffocate. Adult supervision is required.

Specifications

Color White
Height (Metric) 3 mm
Material Plastic
Quantity 10 Pk.
Width (Metric) 3 mm
Product Type Pop Beads