missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ponceau S Staining Solution

Catalog No. A40000278X3
Change view
Click to view available options
Quantity:
1000 mL
3 x 1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40000278X3 3 x 1000 mL
A40000278 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A40000278X3 Supplier Thermo Scientific™ Supplier No. A40000278X3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.

Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein

Specifications

Detection Method Colorimetric
Formulation 0.1% (w/v) in 5% acetic acid
For Use With (Application) Western Blotting
Quantity 3 x 1000 mL
Product Type Ponceau S Staining Solution
What is the best way to image Ponceau staining with an iBright Imaging System?

The recommended imaging method depends on the iBright imager model being used. For iBright FL1500 and CL1500 Imaging Systems, we recommend imaging Ponceau-stained samples with the Ponceau application that can be found in Universal mode. For the iBright CL750 Imaging System, we recommend imaging Ponceau-stained samples in Protein gel mode.

We always recommend imaging with the orange iBright System Contrast Tray (Cat. No. A56597), and if needed, adjust the focus or zoom to get bands as sharp as possible. Better quality pictures are obtained when the membrane is wet.