missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ponceau S, pure

Catalog No. AC161470100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC161470100 10 g
AC161470250 25 g
AC161471000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161470100 Supplier Thermo Scientific Chemicals Supplier No. 161470100

CAS: 6226-79-5 | C22H12N4Na4O13S4 | 760.552 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6226-79-5
Molecular Formula C22H12N4Na4O13S4
Molecular Weight (g/mol) 760.552
MDL Number MFCD00003892
InChI Key VSXKEUCERCWGKF-STNZDNLRSA-J
Synonym C.I. 27195, 3-Hydroxy-4-[2-sulfo-4-(4-sulfophenylazo)phenylazo]-2, 7-naphthalenedisulfonic acid, tetrasodium salt
PubChem CID 11320219
IUPAC Name tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Dye Content 70% min.
Solubility Information Solubility in water: 10g/L. Other solubilities: insoluble in ether
Infrared Spectrum Authentic
Formula Weight 760.54
Grade Pure
Concentration or Composition (by Analyte or Components) Solubility    •To pass testE1%1cm    •≥350L g−1cm−1Lambda Max. (λ)    •514nm±2nm
Packaging Glass bottle
UV Spectrometry Complies
Color Bordeaux to Brown/Red
Quantity 10 g
Physical Form Crystalline Powder
Chemical Name or Material Ponceau S
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-319-2
RTECSNumber QJ6600000
TSCA TSCA
Recommended Storage RT

RUO – Research Use Only