missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ponceau S, MP Biomedicals
$64.24 - $299.64
Chemical Identifiers
| CAS | 6226-79-5 |
|---|---|
| Molecular Formula | C22H12N4Na4O13S4 |
| Molecular Weight (g/mol) | 760.552 |
| InChI Key | VSXKEUCERCWGKF-STNZDNLRSA-J |
| Synonym | 3-Hydroxy-4-[2-sulfo-4-(4-sulfo- phenylazo)phenylazo]-2, 7- naphthalene-disulfonic acid tetrasodium salt |
| PubChem CID | 11320219 |
| IUPAC Name | tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate |
| SMILES | C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15194210
|
MP Biomedicals Inc
0215194210 |
10 g |
N/A
|
|
|||||
|
ICN15194250
|
MP Biomedicals Inc
0215194250 |
50 g |
N/A
|
|
|||||
|
ICN15194280
|
MP Biomedicals Inc
0215194280 |
100 g |
N/A
|
|
|||||
Chemical Identifiers
| 6226-79-5 | |
| 760.552 | |
| 3-Hydroxy-4-[2-sulfo-4-(4-sulfo- phenylazo)phenylazo]-2, 7- naphthalene-disulfonic acid tetrasodium salt | |
| tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate |
| C22H12N4Na4O13S4 | |
| VSXKEUCERCWGKF-STNZDNLRSA-J | |
| 11320219 | |
| C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+] |
Specifications
| 6226-79-5 | |
| C22H12N4Na4O13S4 | |
| VSXKEUCERCWGKF-STNZDNLRSA-J | |
| tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate | |
| 11320219 | |
| 10 g | |
| Ponceau S |
| ∽ 75% | |
| 3-Hydroxy-4-[2-sulfo-4-(4-sulfo- phenylazo)phenylazo]-2, 7- naphthalene-disulfonic acid tetrasodium salt | |
| C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+] | |
| 760.552 | |
| 760.6 | |
| Powder |
Safety and Handling
Recommended Storage : Store at Room Temperature.