missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ponceau S, MP Biomedicals
Specifications
Specifications
| Assay Percent Range | ∽ 75% |
| Quantity | 10 g |
| Physical Form | Powder |
| Formula Weight | 760.6 |
| Chemical Name or Material | Ponceau S |
Safety and Handling
| Recommended Storage | Store at Room Temperature. |