missing translation for 'onlineSavingsMsg'
Learn More

Ponceau S, Electrophoresis Grade

Protein stain for electrophoresis | CAS: 6226-79-5 | C22H12N4Na4O13S4 | 760.552 g/mol

Supplier:  Thermo Scientific Chemicals J6074409

 View more versions of this product

Catalog No. AAJ6074409


Only null left
Explore available promotions

Description

Description

Ponceau S dye is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. It is also suitable for electrophoresis, azo dye for reversible protein staining. This stain can also be used as a fiducial marker in mass spectrometric imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ponceau S dye is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. It is also suitable for electrophoresis, azo dye for reversible protein staining. This stain can also be used as a fiducial marker in mass spectrometric imaging.

Solubility
Soluble in water (10 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
Specifications

Specifications

Ponceau S
C22H12N4Na4O13S4
C.I. 27195
VSXKEUCERCWGKF-STNZDNLRSA-J
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
11320219
Electrophoresis
10 g
6226-79-5
MFCD00003892
Soluble in water (10g/L) at 20°C. Insoluble in ether.
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
760.552
760.58
Brown to Red
Powder

Chemical Identifiers

6226-79-5
760.552
VSXKEUCERCWGKF-STNZDNLRSA-J
11320219
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
C22H12N4Na4O13S4
MFCD00003892
C.I. 27195
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-319-2

RTECSNumber : QJ6600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only