missing translation for 'onlineSavingsMsg'
Learn More

Ponceau 3R, Spectrum™ Chemical

$331.20 - $2645.03

Chemical Identifiers

CAS 3564-09-8
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612428
View Documents
Spectrum Chemical Mfg Cor
PO1445GM
5 g Amber Glass Bottle
Each for $331.20
Only null left
 
18612427
View Documents
Spectrum Chemical Mfg Cor
PO14425GM
25 g Amber Glass Bottle
Each for $1,017.47
Only null left
 
18612426
View Documents
Spectrum Chemical Mfg Cor
PO144100GM
100 g Amber Glass Bottle
Each for $2,645.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3564-09-8

Specifications

3564-09-8
Reagent
5 g
1
Amber Glass Bottle
Ponceau 3R
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.