missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pomalidomide

Chemical Identifiers

CAS 19171-19-8
Molecular Formula C13H11N3O4
Molecular Weight (g/mol) 273.25
MDL Number MFCD12756407
InChI Key UVSMNLNDYGZFPF-UHFFFAOYNA-N
IUPAC Name 4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC465700010
View Documents
Thermo Scientific™
465700010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC465702500
View Documents
Thermo Scientific™
465702500
250 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

19171-19-8
273.25
UVSMNLNDYGZFPF-UHFFFAOYNA-N
NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O
C13H11N3O4
MFCD12756407
4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione

Specifications

19171-19-8
MFCD12756407
UVSMNLNDYGZFPF-UHFFFAOYNA-N
4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
273.24
C13H11N3O4
1 g
NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O
273.25
Pomalidomide
Videos
Safety and Handling

Safety and Handling

GHS H Statement Suspected of damaging fertility or the unborn child.

GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only