missing translation for 'onlineSavingsMsg'
Learn More

GVS™ Polywipe Sponge Ind. Wrapped Pre-Moistened

Catalog No. 10486197
Change view
Click to view available options
Saturation Solution:
neutralizing buffer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Saturation Solution
10486197 neutralizing buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10486197 Supplier GVS™ Supplier No. 10498521

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polywipe is a blue sponge that is premoistened with neutralizing buffer to neutralize the effects of surface disinfectants. For recovery and analysis of microorganisms from a surface.

Description

  • Polywipe is a blue sponge that is premoistened with neutralizing buffer to neutralize the effects of surface disinfectants.

Features and Benefits:

  • Pre-moistened and Sterile.
  • The sponge material is selected to be free of the preservatives found in commercially available sponges, which can inhibit microorganism growth.
  • Polywipe sponges are biocide free and tested for zero toxicity to microorganisms.
  • Each sponge is individually wrapped in a peel pouch and gamma irradiated to ensure sterility.

Application:

  • Used for the recovery and analysis of microorganisms from a surface.
  • Wipe surfaces to be analyzed. Wring fluid onto plate for analysis.
  • Use Sterile Gloves when handling.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Product Type Sponge
Color Blue
Description Polywipe Sponge for microorganism recovery
Format ind. wrapped in peel pouch
Packaging Quantity 50/Pk.
Saturation Solution neutralizing buffer
Sterility Sterile