missing translation for 'onlineSavingsMsg'
Learn More

PolyCo™ PolyWear™ Smooth Disposable Gowns

Catalog No. 18567
Encompass
Change view
Click to view available options
Color:
Blue
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18567 Blue Medium
18568 Blue Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18567 Supplier PolyCo™ Supplier No. GLB 10400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lightweight, single-layer, low-density polyethylene gown for clinical and laboratory settings.

Lightweight, single-layer, low-density polyethylene gown for clinical and laboratory settings

Heat-sealed seam is behind sleeves—minimizes chance of contamination passing through.

  • Full-length wraparound with open back tailored to eliminate excess material
  • Tear-away collar for quick and easy removal
  • Thumb loop on sleeve keeps cuffs under gloves, enhancing leak protection
  • Gowns are folded for quick shake-out and easy donning
  • Powder blue with smooth matte finish

Order Info

Packed individually or bulk in dispenser box.

Compliance

Complies with FDA regulations. Successfully tested for liquid and aerosol blood resistance under ASTM™ F 1670 Synthetic Blood Penetration Resistance and ASTM 1671 Bacteriophage Penetration Resistance.

Specifications

Size Medium
Material Polyethylene Film (Virgin)
Color Blue
Disposable Single-use
Sterility Non-sterile
Finish Smooth Matte
Model 0400
Certifications/Compliance ASTM F 1670, ASTM 1671
Autoclavable Not Autoclavable
Length (Metric) 107 cm
Length (English) 42 in.
For Use With (Application) Clinical, Laboratory
Product Type General Purpose Gown
No. of Pockets None
Gender Unisex
Show More Show Less