missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, M.W. 8,000

Catalog No. AA4162622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4162622 100 g
AA4162636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4162622 Supplier Thermo Scientific Chemicals Supplier No. 04162622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

A cryoprotectant, also useful in plant cell culture research | CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol

A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as a cryoprotectant. It is used as a binder for tablets in pharmaceuticals. It is added to iodine and forms a complex povidone-iodine, which is having disinfectant properties, used in various products like solutions, ointment and pessaries. It is used as an adhesive in glue stick, emulsifier, additive for batteries, ceramics, fiberglass, inks, and inkjet paper, thickening agent in tooth whitening gels and to increase resolution in photoresists for cathode ray tubes. Further, it is used in personal care products such as shampoos, toothpastes, hair sprays and hair gels. In addition to this, it finds application in contact lens solutions, disintegrant for solution polymerization and in steel-quenching solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Physical Form Powder
Melting Point 130°C
Merck Index 14,7697
Solubility Information Soluble in water,alcohol,chlorinated hydrocarbons,amines,nitro paraffins,lower wt fatty acids and chloroform. Insoluble in ether.
Quantity 100 g
Odor Odorless
Chemical Name or Material Polyvinylpyrrolidone, M.W. 8,000
Hazard Category H500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only