Learn More
Polyvinylpyrrolidone, M.W. 40,000
A cryoprotectant, also useful in plant cell culture research | CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
Supplier: Thermo Scientific Chemicals J6241736
Description
A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| White | |
| Powder | |
| (C6H9NO)n | |
| 14,7697 | |
| Soluble in water. | |
| WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
| 1-ethenylpyrrolidin-2-one | |
| 6917 | |
| 40 kd |
| Polyvinylpyrrolidone | |
| 9003-39-8 | |
| MFCD01076626 | |
| n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
| 500 g | |
| *-CC(-*)N1CCCC1=O | |
| 111.14 | |
| CHEBI:82551 |
Chemical Identifiers
| 9003-39-8 | |
| 111.14 | |
| WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
| 6917 | |
| 1-ethenylpyrrolidin-2-one |
| (C6H9NO)n | |
| MFCD01076626 | |
| n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
| CHEBI:82551 | |
| *-CC(-*)N1CCCC1=O |
Safety and Handling
H500
RTECSNumber : TR8370000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only