missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, M.W. 10,000

Catalog No. AAJ60382A1
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60382A1 1 kg
AAJ6038214 25 g
AAJ6038222 100 g
AAJ6038236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ60382A1 Supplier Thermo Scientific Chemicals Supplier No. J60382A1
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cryoprotectant, also useful in plant cell culture research | CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol

Polyvinylpyrrolidone is a polymer used as a pharmaceutical aid, complexing agent, and solubilizer. PVP was used as a plasma volume expander for trauma victims. It acts as diluent, solvent and cleaning agent in various kinds of industries. It also acts as an adjuvant, emulsifier, filtering aid, tableting aid, thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
Physical Form Powder
Merck Index 14,7697
Solubility Information Soluble in water and most organic solvents.
Quantity 1 kg
Formula Weight 10 kd
Chemical Name or Material Polyvinylpyrrolidone
Hazard Category H500
RTECSNumber TR8370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only