missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, M.W. 1,300,000

Catalog No. p-7058992
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4372818 50 g
AA4372830 250 g
AA43728A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4372818 Supplier Thermo Scientific Chemicals Supplier No. 04372818

CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol

Polyvinylpyrrolidone is used to make hair sprays, shampoos, plasma substitutes, detergents and adhesives. It is also used in hair dyes, clarifying agents in wine, beer, and juices, thickeners in inks and paints, water binding agents and protective colloids. It finds application as a special additive for batteries, ceramics, fiberglass, inks, and inkjet, an inhibitor of recrystallisation and to increase resolution in photoresists for cathode ray tubes. Further, it is used for gene delivery. In addition to this, it is used as a pharmaceutical aid, complexing agent and solubilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Physical Form Powder
Merck Index 14,7697
Solubility Information Soluble in water,alcohol,chlorinated hydrocarbons,amines,nitro paraffins,lower wt fatty acids and chloroform. Insoluble in ether.
Quantity 50 g
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Polyvinylpyrrolidone, M.W. 1,300,000
Hazard Category H500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only