missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone K 90 Average Molecular Wt. 360,000, TCI America™

Catalog No. P047325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P047325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P047325G Supplier TCI America Supplier No. P047325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Polyvinylpyrrolidone K 90 Average Molecular Wt. 360,000
EINECSNumber (6)-1007&(6)-1048
RTECSNumber TR8370000
TSCA Yes