missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone K-15, Spectrum™ Chemical

Catalog No. 18605415
Change view
Click to view available options
Quantity:
500 g
5 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605415 500 g Poly Bottle
18605417 100 lb. Fiber Drum
18605416 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605415 Supplier Spectrum Chemical Mfg Cor Supplier No. PO146500GM

PO146, 9003-39-8, (C6H9NO)n

Spectrum™ Chemical Polyvinylpyrrolidone K-15 is also known as polyvidone or povidone. It has been used for trauma victims as a plasma volume expander, as well as a binder in some pharmaceutical tablets. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Grade Reagent