missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylpyrrolidone K 15 Average Molecular Wt. 10,000, TCI America™
$38.51 - $240.64
Chemical Identifiers
| CAS | 9003-39-8 |
|---|---|
| Molecular Formula | (C6H9NO)n |
| Molecular Weight (g/mol) | 111.14 |
| MDL Number | MFCD01076626 |
| InChI Key | WHNWPMSKXPGLAX-UHFFFAOYSA-N |
| Synonym | n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone |
| PubChem CID | 6917 |
| ChEBI | CHEBI:82551 |
| IUPAC Name | 1-ethenylpyrrolidin-2-one |
| SMILES | *-CC(-*)N1CCCC1=O |
Chemical Identifiers
| 9003-39-8 | |
| 111.14 | |
| WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
| 6917 | |
| 1-ethenylpyrrolidin-2-one |
| (C6H9NO)n | |
| MFCD01076626 | |
| n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
| CHEBI:82551 | |
| *-CC(-*)N1CCCC1=O |
Specifications
| 9003-39-8 | |
| (C6H9NO)n | |
| 25 g | |
| WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
| 1-ethenylpyrrolidin-2-one | |
| 6917 | |
| Powder |
| White-Yellow | |
| MFCD01076626 | |
| n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
| *-CC(-*)N1CCCC1=O | |
| 111.14 | |
| CHEBI:82551 | |
| Polyvinylpyrrolidone K 15 Average Molecular Wt. 10,000 |
Safety and Handling
EINECSNumber : (6)-1007&(6)-1048
RTECSNumber : TR8370000
TSCA : Yes