missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylpyrrolidone-iodine complex
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 300°C |
| Color | Brown-Red to Yellow |
| Loss on Drying | 8% max. (5 g, 105°C) |
| Infrared Spectrum | Authentic |
| Linear Formula | (C6H9NO)n·xI |
| Quantity | 2.5 kg |
| Merck Index | 15, 7815 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol,isopropanol,glycerol;,practically insoluble in acetone |
| Percent Purity | 9.0 to 12.0% (available iodine) (on dried substance) |
| Physical Form | Powder |
| Show More |
Safety and Handling
| RTECSNumber | TR1579600 |
| TSCA | TSCA |
RUO – Research Use Only