missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylpyrrolidone, average M.W. 50.000, K30
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| pH | 3.0 to 5.0 (5% aq. soln.) |
| Physical Form | Crystalline Powder or Crystals |
| Melting Point | 225.0°C |
| Avg. Mol. Wt. or Mol. Wt. Range | 50.000 |
| Merck Index | 15, 7814 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether |
| Quantity | 100 g |
| Percent Purity | 11.5 to 12.8% (Nitrogen, on anh. subst.) 10ppm 1-vinylpyrrolidin-2-one (HPLC) max. 3% max. 2-pyrrolidone (HPLC) |
| Identification | B,C,D,E comply |
| Show More |
Safety and Handling
| Hazard Category | HYGROSCOPIC |
| Hazard Statement | LIGHT SENSITIVE |
| RTECSNumber | TR8250000 |
| TSCA | TSCA |
RUO – Research Use Only