missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, average M.W. 3500, K12

Catalog No. AC276142500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276142500 250 g Plastic bottle
AC276140010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC276142500 Supplier Thermo Scientific Chemicals Supplier No. 276142500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White to White-Yellow
pH 3.0 to 5.0 (5% aq. soln.)
Physical Form Powder or Flakes
Merck Index 15, 7814
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether
Quantity 250 g
Percent Purity 11.5 to 12.8% (nitrogen, on anh. subst.), 10ppm 1-vinylpyrrolidin-2-one max. (HPLC), 3.0% max. 2-pyrrolidone (HPLC)
Identification Passes Test
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 5% Max. (K.F.)
Chemical Name or Material Polyvinylpyrrolidone, K12
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement LIGHT SENSITIVE
RTECSNumber TR8250000
TSCA TSCA

RUO – Research Use Only