missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Cream-White to White |
| pH | 4 to 7 (5% aq. soln.) |
| Physical Form | Powder |
| Quantity | 500 g |
| Identification | positive |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Merck Index | 15, 7814 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether |
| Water | 5% max. (K.F.) |
| Show More |
RUO – Research Use Only