missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylformal, TCI America™

Catalog No. P064125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P064125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P064125G Supplier TCI America Supplier No. P064125G
Only null left

Chemical Identifiers

CAS 63450-15-7
Molecular Formula C11H18O5
Molecular Weight (g/mol) 230.26
MDL Number MFCD00084463
InChI Key CCGLZRNKJSJOPC-UHFFFAOYSA-N
PubChem CID 173927
IUPAC Name ethenol;ethenoxymethoxyethene;ethenyl acetate
SMILES CC(=O)OC=C.C=CO.C=COCOC=C

Specifications

Quantity 25 g
Physical Form Granules
Chemical Name or Material Polyvinylformal
EINECSNumber (6)-0716
TSCA Yes