missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Polyurethane Coated Seamless Knit Nylon Gloves

Catalog No. p-7219154
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191304683A Small
191304683B Medium
191304683C Large
191304683D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191304683A Supplier SHOWA™ Supplier No. BO500BS
Only null left

Permeable coating is breathable for comfort, while also protecting the hands from oils and abrasion. Black color is ideal for concealing stains when used in dirty environments.

  • Snug fit maximizes tactile feedback
  • Coating is permeable, allowing hands to breathe to reduce perspiration
  • Light, elastic low-lint glove is resistant to deformation
  • Polyurethane protects the hand from oils and abrasion
  • Seamless construction eliminates irritation
  • No latex allergy risks

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Black, Gray
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Component Assembly, Parts Handling, and HVAC Systems
Glove Cuff Style Knit
Series BO500B
Yarn Gauge 13
Show More Show Less