missing translation for 'onlineSavingsMsg'
Learn More

Polytetrafluoroethylene, 6-10 micron, Thermo Scientific Chemicals

Catalog No. p-7059181
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4418418 50 g
AA4418430 250 g
AA44184A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4418418 Supplier Thermo Scientific Chemicals Supplier No. 04418418
Only null left

Polytetrafluoroethylene, 6-10 micron is used in specially designed coating applications on coil for bakeware.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
SMILES FC(F)(-*)C(F)(F)-*

Specifications

Physical Form Powder
Merck Index 14,7583
Solubility Information Insoluble in water. Soluble in perfluoro kerosene.
Quantity 50 g
Particle Size 6 to 10 Micron
Chemical Name or Material Polytetrafluoroethylene
RTECSNumber KX4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only