missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene standard, M.W. 650,000, Mw/Mn 1.06

Catalog No. AA4194103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4194103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4194103 Supplier Thermo Scientific Chemicals Supplier No. 04194103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-53-6
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name styrene
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Physical Form Powder
Quantity 1 g
Odor Odorless
Chemical Name or Material Polystyrene standard, M.W. 650,000, Mw/Mn 1.06
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only