missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polystyrene Sign: (NO TEXT), White
Improve workplace safety with proper signage and markings. This blank Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$5.29 - $23.32
Specifications
| Material | Polystyrene |
|---|---|
| Shape | Rectangular |
| Displayed Text | Blank |
| Color | White |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|
19104736
|
Brady™
25960 |
7 x 5 in. |
Each for $5.29
|
|
|||||
|
19-104-734
|
Brady™
25376 |
10 x 7 in. |
Each for $13.81
|
|
|||||
|
19104735
|
Brady™
25377 |
14 x 10 in. |
Each for $23.32
|
|
|||||
Description
- Polystyrene material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Polystyrene signs are thin, pliable and durable
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Polystyrene | |
| Rectangular | |
| Blank | |
| White |