missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polystyrene Sign: FALL HAZARD

Improve workplace safety with non-adhesive mounted signage. The Brady™ Fall Hazard Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

Supplier:  Brady™ 132078

 View more versions of this product

Catalog No. 19-106-289

Only null left
Explore available promotions

Description

Description

  • Polystyrene construction is abrasion-resistant and chemical-resistant
  • Corner holes provide easy mounting solutions in a variety of environments
Specifications

Specifications

Not Laminated
1 Ea.
7 in.
-40°F
10 in.
-40°F to 176°F
FALL HAZARD
Polystyrene
Corner Holes
176°F
0.0591 in.
Rectangular
10 x 7 in.
Black on Yellow
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions