missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polystyrene Sign: DO NOT WATCH ARC. VISION DAMAGE MAY OCCUR.
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$13.81 - $20.40
Specifications
| Material | Polystyrene |
|---|---|
| Shape | Rectangular |
| Displayed Text | DO NOT WATCH ARC. VISION DAMAGE MAY OCCUR. |
| Color | Yellow on White |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|
19-112-557
|
Brady™
144244 |
7 x 10 in. |
Each for $13.81
|
|
|||||
|
19-112-558
|
Brady™
144245 |
10 x 14 in. |
Each for $20.40
|
|
|||||
Description
- Polystyrene material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Polystyrene signs are thin, pliable and durable
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Polystyrene | |
| Rectangular | |
| DO NOT WATCH ARC. VISION DAMAGE MAY OCCUR. | |
| Yellow on White |