missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polystyrene Sign: CAUTION - CORROSIVE MATERIALS. WEAR REQUIRED PROTECTION.

Catalog No. 19106449
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
Color:
Black on Yellow
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
19106449 14 x 10 in. Yellow
19106451 10 x 7 in. White
19106860 14 x 10 in. Black on Yellow
19106859 10 x 7 in. Black on Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19106449 Supplier Brady™ Supplier No. 144949

Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Polystyrene
Shape Rectangular
Dimensions (L x W) 14 x 10 in.
Displayed Text CAUTION - CORROSIVE MATERIALS. WEAR REQUIRED PROTECTION.
Color Yellow