Electron Microscopy Sciences Microscope Slide Polystyrene
Catalog No. 50949449
Polystyrene microscope slides are 1" x 3". Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Catalog No.
50949449
Supplier
Electron Microscopy SciencesSupplier No.
71888
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Polystyrene microscope slides are 1" x 3". Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.