Learn More
Corning™ Polystyrene Microplates
GREENER_CHOICE
Designed for in vitro diagnostic assays
$2,841.39
Specifications
Material Polystyrene
View More Specs
Products 1
Catalog Number Mfr. No. Total Volume (Metric) Well Surface Treatment Price Quantity & Availability  
Catalog Number Mfr. No. Total Volume (Metric) Well Surface Treatment Price Quantity & Availability  
07201037
GREENER_CHOICE
View Documents Promotion Details
Corning™
2483
190 μL High Binding
Case of 100 Each for $2,841.39
Only null left
Description
Description
The Stripwell™ low volume microplates allow test kit manufacturers to reduce overall costs, with a reduction in the amount of reagents and antibody necessary to perform assays.
  • Designed with one-half the well area of a 'regular' 8 well strip, providing a much greater surface to volume ratio
  • Recommended working volume of 50μL to 125μL
  • Designed for in vitro diagnostic assays
  • Untreated surface is hydrophobic in nature
  • Color coded strips
  • Bar coding available on request
Maleimide surface covalently couples to sulfhydryl groups (via SH moieties) and are ideal for assays requiring site directed orientation of a particular biomolecule, especially antibodies. Minimal surface preparation is required. Individually packed.
Specifications
Specifications
Polystyrene