Learn More
Greiner Bio-One Polystyrene Microplate Lids
GREENER_CHOICE
Catalog No. 07000286
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
07000286 Sterile
1 options
Catalog No. 07000286 Supplier Greiner Bio-One Supplier No. 656176
For use with non TC-treated 96-well microplates. Greiner Bio-one Polystyrene Microplate Lids is free of detectable DNase, RNase, human DNA.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Height (Metric) 9 mm
Condensation Rings Yes
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.