missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene latex microsphere, 1 micron, 10 wt% dispersion in water

Catalog No. AA419177C
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA419177C 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA419177C Supplier Thermo Scientific Chemicals Supplier No. 0419177C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

Use in protein binding; size markers. The most popular use of latex beads is in the diagnostic use of adsorbed antibodies or antigens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-53-6
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name styrene
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Color White
Physical Form Dispersion
Quantity 15 mL
Odor Odorless
Particle Size 1 Micron
Solubility Information Soluble in Benzene,chloroform,carbon tetrachloride,cyclohexane,dimethylformamide,methylene chloride,pyridine,tetrahydrofuran,toluene,xylene. Insoluble in Acetone,butane,ethanol,ether,methanol,hexane,phenol,propanol,water.
Concentration or Composition (by Analyte or Components) 10 wt% dispersion in water
Chemical Name or Material Polystyrene latex microsphere
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only