missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 60, Spectrum™ Chemical

Catalog No. 18612416
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal.
Packaging:
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612416 500 mL Poly Drum
18612415 4 L Poly Drum
18612414 20 L Poly Drum
18612417 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612416 Supplier Spectrum Chemical Mfg Cor Supplier No. P1822500ML

Chemical Identifiers

CAS 9005-67-8

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Poly Drum
Grade Ungraded
Chemical Name or Material Polysorbate 60

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.