missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 40

Catalog No. AAL15030AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15030AE 100 mL
AAL15030AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15030AE Supplier Thermo Scientific Chemicals Supplier No. L15030AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9005-66-7 | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | 522.68 g/mol

Polysorbate 40 has been used in a study to assess the determination of iodide in urine based on a chemiluminescence system. It has also been used in a study to investigate the delivery of astaxanthin and canthaxanthin to HepG2 cells. Also used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-66-7
Molecular Formula (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165345
InChI Key DVRFSZFBSOKHQW-UHFFFAOYNA-N
Synonym tween r 40, unii-4nr71fn0ia, unii-sti11b5a2x, 4nr71fn0ia, sti11b5a2x, sorbitan, monohexadecanoate, poly oxy-1,2-ethanediyl derivs., polysorbate 40 usan:inn:ban:nf, sorbitan, monopalmitate polyoxyethylene deriv.
PubChem CID 92329579
IUPAC Name 2-[(2R)-2-[(2R,3R,4R)-3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Density 1.09
Quantity 100 mL
Refractive Index 1.47
Solubility Information Fully miscible with water. Soluble in isopropyl alcohol (100mg/ml),and xylene. Insoluble in propylene glycol,and mineral oil.
Chemical Name or Material Polysorbate 40
EINECSNumber 000-000-0
RTECSNumber WG2933000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only