missing translation for 'onlineSavingsMsg'
Learn More

Polyseed™, For BOD, Reagent Grade, LabChem™

Catalog No. lc1888050
Change view
Click to view available options
Quantity:
50 Capsules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC1888050 50 Capsules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC1888050 Supplier LabChem Supplier No. LC1888050
Only null left

Specifications

Chemical Name or Material Polyseed™
Name Note For BOD
Solubility Information Soluble in water
Grade Reagent
Identification Passes Test
Packaging Bottle
Decomposition Information Contains hexavalent Chromium
Quantity 50 Capsules
Physical Form Liquid