Learn More
Eisco™ Polypropylene Volumetric Flask with Screw Cap
Flasks are constructed of high quality, durable polypropylene and fitted with a screw cap.
$35.82 - $35.82
Specifications
Color Clear
Material Polypropylene
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S27345
View Documents
Eisco™
CH0452D
250 mL
Each for $35.82
Only null left
Description
Description
  • Constructed of strong, high quality polypropylene
  • Single, red graduation mark
  • Fitted with a screw cap
  • Autoclavable for easy sterilization
Specifications
Specifications
Clear
Volumetric Flask
Reusable
Narrow
Flat
Polypropylene
Autoclavable
Yes
Non-sterile
Non-treated