missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PPCO Rectangular Carboy with Spigot
GREENER_CHOICE

Catalog No. p-9078034
Change view
Click to view available options
Capacity (Metric):
9 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03007647 9 L
03007648 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03007647
Only null left
Edge

For Use With: Ideal for storing and dispensing lab water and reagents; Autoclavable; Closure Material: PPCO; Color: White

Value your lab can rely on. Fisherbrand PPCO Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.

Ideal for storing and dispensing lab water and reagents

  • Durable construction for long service life
  • Molded graduations for 1 gallon and 51 increments
  • Choice of materials for general use or autoclavability
  • Space-saving rectangular design
  • Wide mouth closure allows for easy filling and cleaning access
  • Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
  • Spigot spout without the adapter fits 5/8-in. ID tubing
  • Materials meet FDA requirements for food use and USP VI criteria (non-toxic)

Specifications

Autoclavable Autoclavable
Capacity (English) 2.4 gal.
Capacity (Metric) 9 L
Certifications/Compliance FDA, USP VI
Closure Material PPCO
Color White
Dimensions (L x W x H) External MaxD (with spigot) x W x H (with closure): 10.5 in. x 6.0 x 14.2 in. (26.7 x 15.3 x 36.1 cm)
For Use With (Application) Ideal for storing and dispensing lab water and reagents
Material PPCO
Mouth Style Wide
Product Type Carboy with Spigot
Shape Rectangular
Show More Show Less