missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Polypropylene Reagent Bottles with Screw Caps
Transparent, rigid polypropylene reagent bottles with screw caps can be used to store liquid solutions in the laboratory. Threaded screw cap renders the bottles spill and leak proof. Autoclavable. Narrow and wide neck varieties available.
Supplier: Eisco™ CH0170BN
Includes: bottle, threaded screw cap
Description
- Made of high quality, rigid polypropylene (PP)
- Includes threaded polypropylene screw cap
- Designed to store liquid solutions in the laboratory
- Spill and leak resistant
- Autoclavable
- Narrow and wide mouth varieties available
Warranty
1 year limited manufacturer warranty
Specifications
| 8 mL | |
| Clear | |
| 1 in. | |
| Screw caps | |
| Polypropylene | |
| 1 Ea. | |
| 25.4 mm | |
| No | |
| 43.2 mm | |
| 1.7 in. | |
| 25.4 mm | |
| Autoclavable |
| 0.27 oz. | |
| Narrow mouth | |
| Solution Storage | |
| 43.18 mm | |
| 25.4 mm | |
| Non-sterile | |
| Yes | |
| Narrow | |
| Reagent Bottle | |
| 13.5 mm | |
| Screw Cap |