Learn More
Eisco™ Polypropylene Laboratory Tray
Eisco™ Polypropylene Laboratory Tray is designed to be rugged, easy to clean and fatigue resistant.
Supplier:  Eisco™ BI0190A
 View more versions of this product
Catalog No. S29354
Only null left
Description
Description
  • Durable polypropylene laboratory trays
  • Chemical and temperature resistant
  • Broad lip for easy carrying
  • Autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
Chemical Identifiers
Autoclavable
Polypropylene
Rectangular
White
1
Specifications
Utility Tray
White
1
54 x 43.5 x 13 cm
54 cm
43.5 cm
13 cm
Autoclavable
Polypropylene
Rectangular
21 in.
17 in.
5 in.
Product Suggestions
Product Suggestions