missing translation for 'onlineSavingsMsg'
Learn More

Tians Polypropylene Lab Coats

Catalog No. p-3203082
Change view
Click to view available options
Size:
Large
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19038440 Large
19038442 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19038440 Supplier Tians Supplier No. 845885L
Only null left

Comfortable with a cloth-like feel

  • Breathable low-lint polypropylene fabric
  • Folded collar
  • Snap front closure
  • Regular style coat features a breast pocket and cut wrists
  • Cleanroom coat style features knit wrists
  • White

Specifications

Product Type Controlled Environments Lab Coat
Size Large
Material Polypropylene
Color White
Cuff Style Open
Disposable Single-use
For Use With (Application) For cleanroom use