missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tians Polypropylene Lab Coats
Description
- Breathable low-lint polypropylene fabric
- Folded collar
- Snap front closure
- Regular style coat features a breast pocket and cut wrists
- Cleanroom coat style features knit wrists
- White
Spécifications
Spécifications
| Product Type | Controlled Environments Lab Coat |
| Size | 2X-Large |
| Material | Polypropylene |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Open |
| Disposable | Single-use |
| For Use With (Application) | For cleanroom use |