missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ Polypropylene Lab Coats, Elastic Wrist, No Pockets
Polypropylene lab coats provide economical, lightweight protection against dirt, grime, and particulates in non-hazardous environments.
Supplier: International Enviroguard™ 2046EXL
Description
- Polypropylene lab coats protect from dust, dirt, grime, and dry particulates in non-hazardous environments
- Often used in food processing, pharmaceuticals, laboratories, cleanrooms, or other controlled environments
- Breathable and lightweight fabric helps keep wearers cool for long hours
- Elastic wrists help keep sleeves in place to prevent skin exposure
- Low-cost protection
Specifications
| Controlled Environments Lab Coat | |
| Unisex | |
| White | |
| Elastic | |
| Non-sterile | |
| None |
| X-Large | |
| Polypropylene | |
| Snaps | |
| Single-use | |
| Particulates | |
| Serged |