missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ Polypropylene Lab Coats, Three Pocket
Polypropylene lab coats feature three convenient pockets and provide economical, lightweight protection against dirt, grime, and particulates in non-hazardous environments.
Supplier: International Enviroguard™ K2045XL
Description
- Polypropylene lab coats protect from dust, dirt, grime, and other dry particulates in non-hazardous environments
- Ideal for laboratories, pharmaceuticals, food processing, environmental cleanup sites, controlled environments
- Breathable fabric helps prevent heat stress and keeps wearers cool over long hours
- Three spacious pockets to carry eyeglasses, pens, and other small instruments
- Low-cost protection
Specifications
| Controlled Environments Lab Coat | |
| Unisex | |
| White | |
| Knit | |
| Non-sterile | |
| 3 | |
| Serged |
| X-Large | |
| Polypropylene | |
| Snaps | |
| Single-use | |
| Particulates | |
| Standard |