missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Cleanroom Compatible Polypropylene Lab Coats

Catalog No. 19813015
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
5X-Large
Closure Type:
Hook and Loop
Snaps
Cuff Style:
Elastic
Knit
No. of Pockets:
None
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Closure Type Cuff Style No. of Pockets
19813015 Small Hook and Loop Elastic None
19168628 Medium Snaps Knit -
19813016 Medium Hook and Loop Elastic None
19168653 - Snaps Knit -
19813017 Large Hook and Loop Elastic None
19813018 X-Large Hook and Loop Elastic None
19168655 - Snaps Knit -
19813019 - Hook and Loop Elastic None
19168656 - Snaps Knit -
19813020 - Hook and Loop Elastic None
19168657 - Snaps Knit -
19168658 5X-Large Snaps Knit -
19813022 - Hook and Loop Elastic None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19813015 Supplier Keystone Safety Supplier No. LC0WENWVSMWHITE

Polypropylene lab coats are made of a breathable fabric that provides economical protection against dry particulates, dirt, and grime.

  • Non-woven 100% Spunbond polypropylene
  • Strong, high tensile strength
  • Lightweight, breathable
  • Cleanroom compatible
  • Low cost solution to protect workers against dry particulates

Specifications

Product Type Controlled Environments Lab Coat
Size Small
Gender Unisex
Color White
Closure Type Hook and Loop
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Latex Free Yes
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Construction Polypropylene
Show More Show Less