missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Polypropylene Graduated Cylinder Protective Collars

Blue Polypropylene Graduated Cylinder Protective Collars feature hexagonal shape to prevent overturned cylinders from rolling off the counter. Graduated cylinders not included.

Supplier:  Eisco™ CH0352D

 View more versions of this product

Catalog No. S88813


Only null left
Explore available promotions

Description

Description

  • Polypropylene protective collars for use with graduated cylinders.
  • Hexagonal shape to prevent an overturned cylinder from rolling of the counter.
  • Multipacks
  • Blue
  • Autoclavable

Warranty

1 year limited manufacturer warranty.

Specifications

Specifications

100 mL
Blue protector
Reusable
100 mL
3.4 oz.
Non-sterile
Hexagonal
Protective Collar
Plastic
Class A, Class B
10 Pk.
Autoclavable
Hexagonal
Protecting 100 mL Graduated Cylinders
25.4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions