missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Polypropylene glycol 1,200

Catalog No. p-7237415
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4736118 50 g
AA4736130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4736118 Supplier Thermo Scientific™ Supplier No. 4736118
Only null left

Specifications

Physical Form Solid
CAS 25322-69-4
Molecular Formula H[OCH(CH{3})CH{2}]+nOH
MDL Number MFCD01779683
Density 1.01
Quantity 50 g