missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polypropylene Disposable Beakers
Disposable or steam-autoclavable for reuse.
Supplier: Customer Channels Group (Ccg) L1220 EMD
Description
- Resistant to most acids, solvents and other common reagents
- Special “dripless” lip
- Rib construction permits stacking without binding
- Graduated markings in mL and oz.
Chemical Identifiers
| Polypropylene | |
| Beaker | |
| Single-use |
| Spout | |
| Autoclavable | |
| Yes |
Specifications
| 50 mL | |
| Spout | |
| Beaker | |
| Single-use |
| Polypropylene | |
| 5 mL | |
| Autoclavable | |
| Yes |