missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ Polypropylene Coveralls with Elastic Wrists and Open Ankles

Catalog No. 17184234
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17184234 Medium
17184235 Large
17184233 X-Large
17184239 2X-Large
17184238 3X-Large
17184237 4X-Large
17184236 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17184234 Supplier International Enviroguard™ Supplier No. 2012M

Disposable coveralls provide low-cost protection against dust, dirt, and grime.

  • Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
  • Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
  • Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
  • Elastic wrists, open ankles
  • Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
  • Stay cool, comfortable, and protected from head-to-toe
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more

Specifications

Product Type Light-duty Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Open
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Dust
Collar Type Fold-down
Latex Free Yes
Model 2012
No. of Pockets None
For Use With (Application) Food Packaging, Food Processing, Warehousing, Laboratories
Show More Show Less