missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Polypropylene Carboy with Silicone-Lined Cap and Spigot
Description
- Durable polypropylene design: Made from virgin-grade polypropylene with a silicone-lined cap and integrated spigot for easy dispensing, ideal for liquid storage and transportation.
- Autoclavable up to 135°C: Withstands steam sterilization and temperature ranges from 0-135°C, making it suitable for scientific and lab applications.
- Certified for safety: FDA 21 CFR 177.1520 and USP 88 Class VI compliant, ensuring safe use in food, drug, and medical applications.
- Leakproof tested: Rigorously tested for crack resistance, handle strength, and leak prevention, ensuring secure and worry-free liquid handling.
- Versatile applications: Perfect for reagent storage, liquid mixing, and dispensing in labs, research facilities, industrial environments, and educational settings.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 25 L |
| Color | Translucent |
| Material | Polypropylene |
| Mouth Style | Narrow |
| Product Type | Carboy |
| Sterility | Non-sterile |